1-[4-(2-Phenylethyl)benzyl]naphthalene ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
1-[4-(2-Phenethyl)benzyl]naphthalene
P1181-1G
P1181-25G
TCP1181-001GEA
116.92
CAD
TCP1181-001G
TCP1181-25G
1-[4-(2-Phenylethyl)benzyl]naphthalene ≥98.0% (by HPLC)
1-[4-(2-Phenylethyl)benzyl]naphthalene
CAS Number: 127833-53-8
MDL Number: MFCD00191572
Molecular Formula: C25H22
Molecular Weight: 322.45
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 66
Other Product Names / Synonyms: 1-[4-(2-Phenethyl)benzyl]naphthalene
Formula:
C₂₅H₂₂ MW: 322.45 g/mol Melting Pt: 66 °C |
MDL Number:
MFCD00191572 CAS Number: 127833-53-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...