(±)-1,2-Pentanediol ≥98.0%

Supplier: TCI America

Synonyms: (±)-1,2-Dihydroxy pentane, 1,2-Pentanediol

P1178-500ML P1178-25ML
TCP1178-500MLEA 719.71 CAD
TCP1178-500ML TCP1178-025ML
(±)-1,2-Pentanediol ≥98.0%
(±)-1,2-Pentanediol

CAS Number: 5343-92-0
MDL Number: MFCD00010736
Molecular Formula: C5H12O2
Molecular Weight: 104.15
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 210
Flash Point (°C): 104
Specific Gravity (20/20): 0.97


Other Product Names / Synonyms: 1,2-Dihydroxypentane

Formula: C₅H₁₂O₂
MW: 104.15 g/mol
Boiling Pt: 210 °C
Density: 0.97 (20 °C)
Flash Pt: 104 °C
Storage Temperature: Ambient
MDL Number: MFCD00010736
CAS Number: 5343-92-0
EINECS: 226-285-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bb275f85c973f605c8d0e05f8f20d7ebb3519157&vid=e4467ef18791111ad495c7a007c9d4ffec2e6f0c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR