Tri-p-tolyl phosphite
Supplier: TCI America
P1175-25G
P1175-500G
TCP1175-025GEA
69.36
CAD
TCP1175-025G
TCP1175-500G
Tri-p-tolyl phosphite
Tri-p-tolyl phosphite
CAS Number: 620-42-8
MDL Number: MFCD00041909
Molecular Formula: C21H21O3P
Molecular Weight: 352.37
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 217
Specific Gravity (20/20): 1.13
Other Product Names / Synonyms: Phosphorous Acid Tri-p-cresyl Ester, Phosphorous Acid Tris(4-methylphenyl) Ester, Phosphorous Acid Tri-p-tolyl Ester, Tri-p-cresyl Phosphite, Tris(4-methylphenyl) Phosphite
Formula:
C₂₁H₂₁O₃P Density: 1.13 (20 °C) Flash Pt: 217 °C |
MDL Number:
MFCD00041909 CAS Number: 620-42-8 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...