trans-2-Phenylcyclohexanol ≥98.0%
Supplier: TCI America
P1174-1G
P1174-5G
TCP1174-001GEA
131.14
CAD
TCP1174-001G
TCP1174-5G
trans-2-Phenylcyclohexanol ≥98.0%
trans-2-Phenylcyclohexanol
CAS Number: 2362-61-0
MDL Number: MFCD00003865
Molecular Formula: C12H16O
Molecular Weight: 176.26
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 122
Melting point (°C): 58
Flash Point (°C): 113
Formula:
C₁₂H₁₆O |
MDL Number:
MFCD00003865 CAS Number: 2362-61-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...