Tris(2-ethylhexyl)phosphite ≥90.0%
Supplier: TCI America
Synonyms:
Phosphorous Acid Tris(2-ethylhexyl) Ester, Phosphorous Acid Trioctyl Ester, Trioctyl Phosphite
P1169-25ML
P1169-500ML
TCP1169-025MLEA
67.53
CAD
TCP1169-025ML
TCP1169-500ML
Tris(2-ethylhexyl)phosphite ≥90.0%
Tris(2-ethylhexyl)phosphite
CAS Number: 301-13-3
MDL Number: MFCD00015266
Molecular Formula: C24H51O3P
Molecular Weight: 418.64
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 194
Specific Gravity (20/20): 0.89
Other Product Names / Synonyms: Phosphorous Acid Trioctyl Ester, Phosphorous Acid Tris(2-ethylhexyl) Ester, Trioctyl Phosphite
Formula:
C₂₄H₅₁O₃P MW: 418.64 g/mol Density: 0.89 (20 °C) Flash Pt: 194 °C |
MDL Number:
MFCD00015266 CAS Number: 301-13-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...