Pentaethylene glycol monomethyl ether ≥96.0%
Supplier: TCI America
P1159-1G
P1159-5G
TCP1159-001GEA
261.17
CAD
TCP1159-001G
TCP1159-005G
Pentaethylene glycol monomethyl ether ≥96.0%
Pentaethylene glycol monomethyl ether
CAS Number: 23778-52-1
MDL Number: MFCD00045995
Molecular Formula: C11H24O6
Molecular Weight: 252.31
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 1.08
Storage Temperature: 0-10°C
Product Categories: Bioscience, PEGylation Reagents
Other Product Names / Synonyms: mPEG5-Alcohol
Formula:
C₁₁H₂₄O₆ Density: 1.08 (20 °C) |
MDL Number:
MFCD00045995 CAS Number: 23778-52-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...