3-(3-Pyridyl)acrylic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
P1157-25G
P1157-5G
TCP1157-025GEA
175.22
CAD
TCP1157-025G
TCP1157-005G
3-(3-Pyridyl)acrylic acid ≥98.0% (by titrimetric analysis)
3-(3-Pyridyl)acrylic acid
CAS Number: 1126-74-5
MDL Number: MFCD00006410
Molecular Formula: C8H7NO2
Molecular Weight: 149.15
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 234
Other Product Names / Synonyms: 3-Pyridineacrylic Acid
Formula:
C₈H₇NO₂ Melting Pt: 234 °C |
MDL Number:
MFCD00006410 CAS Number: 1126-74-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...