2-Allenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥95.0% (by GC, titration analysis)

Supplier: TCI America

A2086-5G A2086-1G
TCA2086-5GEA 636.42 CAD
TCA2086-5G TCA2086-1G
2-Allenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥95.0% (by GC, titration analysis)
2-Allenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

CAS Number: 865350-17-0
MDL Number: MFCD09265121
Molecular Formula: C9H15BO2
Molecular Weight: 166.03
Purity/Analysis Method: >95.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 81
Flash Point (°C): 66
Specific Gravity (20/20): 0.94
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Boronic Acids & Boronic Acid Derivatives
Other Product Names / Synonyms: Allenylboronic Acid Pinacol Ester

Formula: C₉H₁₅BO₂
Boiling Pt: 81 °C
Density: 0.94 (20 °C)
Flash Pt: 66 °C
MDL Number: MFCD09265121
CAS Number: 865350-17-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8af1ee744dcf9e98766a29993feac6f9f9305ee1&vid=8af1ee744dcf9e98766a29993feac6f9f9305ee1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR