N-Phenylaza-15-crown-5 ≥96.0%

Supplier: TCI America

Synonyms: 13-Phenyl-1,4,7,10-tetraoxa-13-azacyclopentadecane, N-Phenyl-13-aza-1,4,7,10-tetraoxacyclopentadecane, N-Phenylaza-15-crown 5-Ether

P1143-1G P1143-5G
TCP1143-001GEA 159.58 CAD
TCP1143-001G TCP1143-005G
N-Phenylaza-15-crown-5 ≥96.0%
N-Phenylaza-15-crown-5

CAS Number: 66750-10-5
MDL Number: MFCD00040513
Molecular Formula: C16H25NO4
Molecular Weight: 295.38
Purity/Analysis Method: >96.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 46


Product Categories: Materials Chemistry, Macrocycles for Host-Guest Chemistry
Other Product Names / Synonyms: N-Phenyl-13-aza-1,4,7,10-tetraoxacyclopentadecane

Formula: C₁₆H₂₅NO₄
MW: 295.37 g/mol
Melting Pt: 46 °C
MDL Number: MFCD00040513
CAS Number: 66750-10-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eb7c769ee973ea79ccfd3600f530568c45dcec6b&vid=eb7c769ee973ea79ccfd3600f530568c45dcec6b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR