(R)-3-Hydroxypyrrolidine hydrochloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

P1140-5G P1140-25G
TCP1140-005GEA 150.42 CAD
TCP1140-005G TCP1140-025G
(R)-3-Hydroxypyrrolidine hydrochloride ≥98.0% (by titrimetric analysis)
(R)-3-Hydroxypyrrolidine hydrochloride

CAS Number: 104706-47-0
MDL Number: MFCD00191570
Molecular Formula: C4H9NO
Molecular Weight: 123.58
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 109
Specific rotation [a]20/D: -7.8 deg (C=3.5, MeOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (R)-(-)-3-Hydroxypyrrolidine Hydrochloride

Formula: C₄H₁₀ClNO
Melting Pt: 109 °C
MDL Number: MFCD00191570
CAS Number: 104706-47-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ffe8d341f961654913372a1a4001e32eea1e69ae&vid=ffe8d341f961654913372a1a4001e32eea1e69ae&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR