Monobutyl phthalate ≥95.0%

Supplier: TCI America

Synonyms: 2-(Butoxycarbonyl)benzoic acid, Butyl Hydrogen Phthalate, Phthalic Acid Monobutyl Ester

P1132-5G P1132-25G
TCP1132-5GEA 131.31 CAD
TCP1132-5G TCP1132-025G
Monobutyl phthalate ≥95.0%
Monobutyl phthalate

CAS Number: 131-70-4
MDL Number: MFCD00043498
Molecular Formula: C12H14O4
Molecular Weight: 222.24
Purity/Analysis Method: >95.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 73


Other Product Names / Synonyms: Phthalic Acid Monobutyl Ester, Butyl Hydrogen Phthalate

Formula: C₁₂H₁₄O₄
MW: 222.24 g/mol
Melting Pt: 73 °C
MDL Number: MFCD00043498
CAS Number: 131-70-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=04dff32cdce6512945fbce6487a8dcb0d322b719&vid=04dff32cdce6512945fbce6487a8dcb0d322b719&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR