3-Phenylthiophene ≥95.0%
Supplier: TCI America
P1126-5G
P1126-1G
TCP1126-5GEA
560.9
CAD
TCP1126-5G
TCP1126-001G
3-Phenylthiophene ≥95.0%
3-Phenylthiophene
CAS Number: 2404-87-7
MDL Number: MFCD00114997
Molecular Formula: C10H8S
Molecular Weight: 160.23
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 256
Melting point (°C): 91
Formula:
C₁₀H₈S Boiling Pt: 256 °C Melting Pt: 91 °C |
MDL Number:
MFCD00114997 CAS Number: 2404-87-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...