(S)-1-Phenyl-2-(p-tolyl)ethylamine ≥98.0%
Supplier: TCI America
P1118-25G
P1118-5G
TCP1118-025GEA
1363.54
CAD
TCP1118-025G
TCP1118-5G
(S)-1-Phenyl-2-(p-tolyl)ethylamine ≥98.0%
(S)-1-Phenyl-2-(p-tolyl)ethylamine
CAS Number: 30339-30-1
MDL Number: MFCD00221505
Molecular Formula: C15H17N
Molecular Weight: 211.31
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 164
Specific Gravity (20/20): 1.02
Specific rotation [a]20/D: 11 deg (neat)
Formula:
C₁₅H₁₇N Density: 1.02 (20 °C) Flash Pt: 164 °C |
MDL Number:
MFCD00221505 CAS Number: 30339-30-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...