1,3-Diaminopropane-N,N'-diacetic acid ≥98.0%

Supplier: TCI America

Synonyms: 1,3-Propylenediamine-N,N'-diacetic Acid

P1115-5G P1115-25G
TCP1115-005GEA 52.14 CAD
TCP1115-005G TCP1115-025G
1,3-Diaminopropane-N,N'-diacetic acid ≥98.0%
1,3-Diaminopropane-N,N'-diacetic acid

CAS Number: 112041-05-1
MDL Number: MFCD00142534
Molecular Formula: C7H14N2O4
Molecular Weight: 190.20
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White


Other Product Names / Synonyms: 1,3-Propylenediamine-N,N'-diacetic Acid

Formula: C₇H₁₄N₂O₄
MDL Number: MFCD00142534
CAS Number: 112041-05-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d25e8b5fff1ed1381a53de76d57fa68712d2d1a0&vid=d25e8b5fff1ed1381a53de76d57fa68712d2d1a0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR