2-Phenoxypropanol 96,0

Supplier: TCI America

Synonyms: Propylene Glycol 2-Monophenyl Ether

P1111-5G P1111-25G
TCP1111-5GEA 183.28 CAD
TCP1111-5G TCP1111-25G
2-Phenoxypropanol 96,0
2-Phenoxypropanol

CAS Number: 4169-04-4
MDL Number: MFCD00142958
Molecular Formula: C9H12O2
Molecular Weight: 152.19
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 244
Specific Gravity (20/20): 1.07


Other Product Names / Synonyms: Propylene Glycol 2-Monophenyl Ether

Formula: C₉H₁₂O₂
MW: 152.19 g/mol
Boiling Pt: 244 °C
Density: 1.07 (20 °C)
MDL Number: MFCD00142958
CAS Number: 4169-04-4

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR