1-(2-Pyrimidyl)piperazine ≥98.0%

Supplier: TCI America

P1110-25G P1110-5G
TCP1110-025GEA 110.6 CAD
TCP1110-025G TCP1110-5G
1-(2-Pyrimidyl)piperazine ≥98.0%
1-(2-Pyrimidyl)piperazine

CAS Number: 20980-22-7
MDL Number: MFCD00040742
Molecular Formula: C8H12N4
Molecular Weight: 164.21
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 120
Melting point (°C): 33
Flash Point (°C): 110


Other Product Names / Synonyms: 2-(1-Piperazinyl)pyrimidine

Formula: C₈H₁₂N₄
Boiling Pt: 120 °C
Melting Pt: 33 °C
Flash Pt: 110 °C
MDL Number: MFCD00040742
CAS Number: 20980-22-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6a87028a8c36270aee536dc2ffc051b0c8808393&vid=6a87028a8c36270aee536dc2ffc051b0c8808393&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR