4-Propenylveratrole ≥98.0% for synthesis
Supplier: TCI America
Synonyms:
4-(1-Propenyl)pyrocatechol dimethyl ether, 4-(1-Propenyl)-1,2-dimethoxybenzene, 1,2-Dimethoxy-4-Propenylbenzene, 1,2-Dimethoxy-4-(1-propenyl)benzene, Methyl isoeugenol, O-Methyl Isoeugenol
CAS Number: 93-16-3
MDL Number: MFCD00009282
Molecular Formula: C11H14O2
Molecular Weight: 178.23
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 264
Flash Point (°C): 100
Specific Gravity (20/20): 1.05
Product Categories: Bioscience, Phenylpropanoids
Other Product Names / Synonyms: 1,2-Dimethoxy-4-(1-propenyl)benzene, O-Methyl Isoeugenol, 4-(1-Propenyl)pyrocatechol Dimethyl Ether
Formula:
C₁₁H₁₄O₂ Boiling Pt: 264 °C Density: 1.05 (20 °C) Flash Pt: 100 °C |
MDL Number:
MFCD00009282 CAS Number: 93-16-3 EINECS: 202-224-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...