9-Phenanthreneboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
P1093-1G
P1093-5G
TCP1093-1GEA
37.92
CAD
TCP1093-1G
TCP1093-5G
9-Phenanthreneboronic acid (contains varying amounts of Anhydride)
9-Phenanthreneboronic acid
CAS Number: 68572-87-2
MDL Number: MFCD00143524
Molecular Formula: C14H11BO2
Molecular Weight: 222.05
Form: Crystal
Color: White
Formula:
C₁₄H₁₁BO₂ MW: 222.05 g/mol |
MDL Number:
MFCD00143524 CAS Number: 68572-87-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...