DL-3-Pyrrolidinol ≥97.0% (by titrimetric analysis)
Supplier: TCI America
P1083-5G
P1083-25G
TCP1083-005GEA
157.05
CAD
TCP1083-005G
TCP1083-025G
DL-3-Pyrrolidinol ≥97.0% (by titrimetric analysis)
DL-3-Pyrrolidinol
CAS Number: 40499-83-0
MDL Number: MFCD00005256
Molecular Formula: C4H9NO
Molecular Weight: 87.12
Purity/Analysis Method: >97.0% (T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 179
Flash Point (°C): 110
Specific Gravity (20/20): 1.08
Other Product Names / Synonyms: DL-3-Hydroxypyrrolidine
Formula:
C₄H₉NO Boiling Pt: 179 °C Density: 1.08 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00005256 CAS Number: 40499-83-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...