3-Amino-1-adamantanol hydrate ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 1-Amino-3-hydroxyadamantane, 3-Aminoadamantan-1-ol

A2079-1G A2079-5G
TCA2079-1GEA 28.44 CAD
TCA2079-1G TCA2079-5G
3-Amino-1-adamantanol hydrate ≥98.0% (by GC, titration analysis)
3-Amino-1-adamantanol hydrate

CAS Number: 702-82-9
MDL Number: MFCD01821204
Molecular Formula: C10H17NO
Molecular Weight: 167.25
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 269
Flash Point (°C): 110


Other Product Names / Synonyms: 1-Amino-3-hydroxyadamantane

Formula: C₁₀H₁₇NO·xH₂O
MW: 167.25 g/mol (anhy)
Melting Pt: 269 °C
Flash Pt: 110 °C
MDL Number: MFCD01821204
CAS Number: 702-82-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6da9a0769b06e696c708e4e39c7a9659ddbb30f&vid=5b682a85fa5f6684630c0404f2a9999dedd0214a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR