Piperonylonitrile ≥98.0%
Supplier: TCI America
P1074-10G
P1074-25G
TCP1074-010GEA
74.42
CAD
TCP1074-010G
TCP1074-025G
Piperonylonitrile ≥98.0%
Piperonylonitrile
CAS Number: 4421-09-4
MDL Number: MFCD00005820
Molecular Formula: C8H5NO2
Molecular Weight: 147.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 94
Other Product Names / Synonyms: 1,3-Benzodioxole-5-carbonitrile, 3,4-Methylenedioxybenzonitrile
Formula:
C₈H₅NO₂ Melting Pt: 94 °C |
MDL Number:
MFCD00005820 CAS Number: 4421-09-4 UN: 3439 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...