4-Propoxyphthalonitrile ≥95.0%

Supplier: TCI America

Synonyms: 1,2-Dicyano-4-propoxybenzene

P1070-1G P1070-5G
TCP1070-001GEA 170.32 CAD
TCP1070-001G TCP1070-005G
4-Propoxyphthalonitrile ≥95.0%
4-Propoxyphthalonitrile

CAS Number: 106144-18-7
MDL Number: MFCD00059102
Molecular Formula: C11H10N2O
Molecular Weight: 186.21
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Color: White


Product Categories: Materials Chemistry, Building Blocks for Phthalocyanines
Other Product Names / Synonyms: 1,2-Dicyano-4-propoxybenzene

Formula: C₁₁H₁₀N₂O
MW: 186.21 g/mol
MDL Number: MFCD00059102
CAS Number: 106144-18-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5dafe77bc7c851d4835b554b0ff77ad2b36c818c&vid=5dafe77bc7c851d4835b554b0ff77ad2b36c818c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR