Pamoic acid disodium salt monohydrate ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 4,4'-Methylenebis(3-hydroxy-2-naphthoic Acid Sodium Salt), Embonic Acid Disodium Salt

P1059-25G P1059-250G
TCP1059-025GEA 97.96 CAD
TCP1059-025G TCP1059-250G
Pamoic acid disodium salt monohydrate ≥98.0% (by HPLC)
Pamoic acid disodium salt monohydrate

CAS Number: 6640-22-8
MDL Number: MFCD00036183
Molecular Formula: C23H16O6
Molecular Weight: 432.34
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: Pale Yellow


Other Product Names / Synonyms: Embonic Acid Disodium Salt, 4,4'-Methylenebis(3-hydroxy-2-naphthoic Acid Sodium Salt)

Formula: C₂₃H₁₆O₆
MW: 432.34 g/mol
MDL Number: MFCD00036183
CAS Number: 6640-22-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7378473f4d92a6739726f1bdaf6fa876ebd3a19e&vid=7378473f4d92a6739726f1bdaf6fa876ebd3a19e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR