10,12-Pentacosadiyn-1-ol ≥98.0%

Supplier: TCI America

P1053-100MG P1053-1G
TCP1053-100MGEA 261.17 CAD
TCP1053-100MG TCP1053-1G
10,12-Pentacosadiyn-1-ol ≥98.0%
10,12-Pentacosadiyn-1-ol

CAS Number: 92266-90-5
MDL Number: MFCD00059096
Molecular Formula: C25H44O
Molecular Weight: 360.63
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 60

Formula: C₂₅H₄₄O
MW: 360.63 g/mol
Melting Pt: 60 °C
MDL Number: MFCD00059096
CAS Number: 92266-90-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f07289517637bfd4b1327fe9c3c5434626e69a9f&vid=f07289517637bfd4b1327fe9c3c5434626e69a9f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR