3,5-Pyrazoledicarboxylic acid monohydrate ≥98.0%
Supplier: TCI America
Synonyms:
1H-Pyrazole-3,5-dicarboxylic acid hydrate, Pyrazole-3,5-dicarboxylic acid monohydrate, 1H-Pyrazole-3,5-dicarboxylic acid monohydrate
P1048-5G
P1048-25G
TCP1048-005GEA
159.58
CAD
TCP1048-005G
TCP1048-025G
3,5-Pyrazoledicarboxylic acid monohydrate ≥98.0%
3,5-Pyrazoledicarboxylic acid monohydrate
CAS Number: 3112-31-0
MDL Number: MFCD00005235
Molecular Formula: C5H4N2O4
Molecular Weight: 156.10
Purity/Analysis Method:
Form: Crystal
Color: White
Melting point (°C): 295
Formula:
C5H4N2O4 MW: 174.11 g/mol Melting Pt: 295 °C |
MDL Number:
MFCD00149323 CAS Number: 3112-31-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...