2,2-Pentamethylene-1,3-dioxolan-4-one ≥98.0%
Supplier: TCI America
Synonyms:
1,4-Dioxaspiro[4.5]decan-2-one, 1,4-Dioxaspiro[4.5]-2-decanone
P1019-5G
P1019-25G
TCP1019-005GEA
88.48
CAD
TCP1019-005G
TCP1019-25G
2,2-Pentamethylene-1,3-dioxolan-4-one ≥98.0%
2,2-Pentamethylene-1,3-dioxolan-4-one
CAS Number: 4423-79-4
MDL Number: MFCD00010852
Molecular Formula: C8H12O3
Molecular Weight: 156.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Flash Point (°C): 105
Freezing point (°C): 31
Other Product Names / Synonyms: 1,4-Dioxaspiro[4.5]-2-decanone
Formula:
C₈H₁₂O₃ Flash Pt: 105 °C |
MDL Number:
MFCD00010852 CAS Number: 4423-79-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...