Didecyl phosphate ≥95.0% (by titrimetric analysis)

Supplier: TCI America

P1011-5G P1011-100MG P1011-1G
TCP1011-5GEA 1044.38 CAD
TCP1011-5G TCP1011-100MG TCP1011-001G
Didecyl phosphate ≥95.0% (by titrimetric analysis)
Didecyl phosphate

CAS Number: 7795-87-1
MDL Number: MFCD00015062
Molecular Formula: C20H43O4P
Molecular Weight: 378.53
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Color: White
Melting point (°C): 47


Other Product Names / Synonyms: Phosphoric Acid Didecyl Ester

Formula: C₂₀H₄₃O₄P
Melting Pt: 47 °C
MDL Number: MFCD00015062
CAS Number: 7795-87-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8b2e826e33df7ce8f868f4c7a3ad4c5ed3be02f9&vid=8b2e826e33df7ce8f868f4c7a3ad4c5ed3be02f9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR