Methyl-N-phenylcarbamate ≥98.0%

Supplier: TCI America

P1010-500G P1010-25G
TCP1010-500GEA 630.42 CAD
TCP1010-500G TCP1010-025G
Methyl-N-phenylcarbamate ≥98.0%
Methyl-N-phenylcarbamate

CAS Number: 2603-10-3
MDL Number: MFCD00027552
Molecular Formula: C8H9NO2
Molecular Weight: 151.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 126
Melting point (°C): 47


Other Product Names / Synonyms: N-Phenylcarbamic Acid Methyl Ester

Formula: C₈H₉NO₂
Boiling Pt: 126 °C
Melting Pt: 47 °C
MDL Number: MFCD00027552
CAS Number: 2603-10-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ee7fed473238433f336daf0b1f4875203a0c6d2&vid=0ee7fed473238433f336daf0b1f4875203a0c6d2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR