4-Piperidineethanol ≥96.0%

Supplier: TCI America

Synonyms: 4-(2-Hydroxyethyl)piperidine, 2-(Piperidin-4-yl)ethanol

P0992-25G P0992-5G
TCP0992-25GEA 447.14 CAD
TCP0992-25G TCP0992-005G
4-Piperidineethanol ≥96.0%
4-Piperidineethanol

CAS Number: 622-26-4
MDL Number: MFCD00006008
Molecular Formula: C7H15NO
Molecular Weight: 129.20
Purity/Analysis Method: >96.0% (GC,T)
Form: Crystal
Color: White
Boiling point (°C): 228
Melting point (°C): 50
Flash Point (°C): 110


Other Product Names / Synonyms: 4-(2-Hydroxyethyl)piperidine

Formula: C₇H₁₅NO
MW: 129.20 g/mol
Boiling Pt: 228 °C
Melting Pt: 50 °C
Flash Pt: 110 °C
MDL Number: MFCD00006008
CAS Number: 622-26-4

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR