Sodium isonicotinate ≥99.0%
Supplier: TCI America
Synonyms:
Isonicotinic acid sodium salt, Sodium Isonicotinate, Pyridine-4-carboxylic acid sodium salt, Sodium Pyridine-4-carboxylate
P0991-25G
P0991-250G
TCP0991-025GEA
116.92
CAD
TCP0991-025G
TCP0991-250G
Sodium isonicotinate ≥99.0%
Sodium isonicotinate
CAS Number: 16887-79-9
MDL Number: MFCD00064868
Molecular Formula: C6H5NO2
Molecular Weight: 145.09
Purity/Analysis Method: >99.0% (HPLC,T)
Form: Crystal
Color: White
Other Product Names / Synonyms: Isonicotinic Acid Sodium Salt, Pyridine-4-carboxylic Acid Sodium Salt, Sodium Pyridine-4-carboxylate
Formula:
C₆H₄NNaO₂ MW: 145.0912 g/mol |
MDL Number:
MFCD00064868 CAS Number: 16887-79-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...