2,6-Pyridinedicarboxaldehyde ≥98.0%
Supplier: TCI America
P0949-1G
P0949-5G
TCP0949-001GEA
222.78
CAD
TCP0949-001G
TCP0949-005G
2,6-Pyridinedicarboxaldehyde ≥98.0%
2,6-Pyridinedicarboxaldehyde
CAS Number: 5431-44-7
MDL Number: MFCD00010103
Molecular Formula: C7H5NO2
Molecular Weight: 135.12
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 122
Formula:
C₇H₅NO₂ Melting Pt: 122 °C |
MDL Number:
MFCD00010103 CAS Number: 5431-44-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...