4-Amino-N-methylbenzamide ≥98.0% (by GC, titration analysis)
Supplier: TCI America
A2064-5G
A2064-1G
TCA2064-5GEA
37.92
CAD
TCA2064-5G
TCA2064-1G
4-Amino-N-methylbenzamide ≥98.0% (by GC, titration analysis)
4-Amino-N-methylbenzamide
CAS Number: 6274-22-2
Molecular Formula: C8H10N2O
Molecular Weight: 150.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 181
Formula:
C₈H₁₀N₂O Melting Pt: 181 °C |
MDL Number:
MFCD00665792 CAS Number: 6274-22-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...