Bis(cis-3,3,5-trimethylcyclohexyl)phthalate ≥98.0%
Supplier: TCI America
P0946-25G
P0946-5G
TCP0946-025GEA
267.02
CAD
TCP0946-025G
TCP0946-5G
Bis(cis-3,3,5-trimethylcyclohexyl)phthalate ≥98.0%
Bis(cis-3,3,5-trimethylcyclohexyl)phthalate
CAS Number: 245652-81-7
MDL Number: MFCD00070480
Molecular Formula: C26H38O4
Molecular Weight: 414.59
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 93
Other Product Names / Synonyms: Phthalic Acid Bis(cis-3,3,5-trimethylcyclohexyl) Ester
Formula:
C₂₆H₃₈O₄ Melting Pt: 93 °C |
MDL Number:
MFCD00070480 CAS Number: 245652-81-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...