trans-4-Propylcyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

P0943-25G P0943-5G
TCP0943-025GEA 129.56 CAD
TCP0943-025G TCP0943-005G
trans-4-Propylcyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)
trans-4-Propylcyclohexanecarboxylic acid

CAS Number: 38289-27-9
MDL Number: MFCD00059559
Molecular Formula: C10H18O2
Molecular Weight: 170.25
Purity/Analysis Method: >98.0% (T,GC)
Form: Crystal
Color: White
Melting point (°C): 98

Formula: C₁₀H₁₈O₂
MW: 170.24 g/mol
Melting Pt: 98 °C
MDL Number: MFCD00059559
CAS Number: 38289-27-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fdedea00a168b3465625c5ec33dcb06f32c4971d&vid=fdedea00a168b3465625c5ec33dcb06f32c4971d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T,GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR