Pyridinium-3-nitrobenzenesulfonate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
P0941-5G
P0941-25G
TCP0941-005GEA
72.68
CAD
TCP0941-005G
TCP0941-025G
Pyridinium-3-nitrobenzenesulfonate ≥98.0% (by titrimetric analysis)
Pyridinium-3-nitrobenzenesulfonate
CAS Number: 84752-61-4
MDL Number: MFCD00060234
Molecular Formula: C11H10N2O5S
Molecular Weight: 282.27
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 124
Formula:
C₁₁H₁₀N₂O₅S MW: 282.27 g/mol Melting Pt: 124 °C |
MDL Number:
MFCD00060234 CAS Number: 84752-61-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...