Pentaerythritol tetrakis [3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate] ≥95.0%

Supplier: TCI America

Synonyms: Tetrakis[methylene-3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate]methane, Antioxidant 1010, 2,2-Bis(((3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoyl)oxy)methyl)propane-1,3-diyl bis(3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoate)

P0932-25G P0932-500G P0932-100G
TCP0932-025GEA 63.06 CAD
TCP0932-025G TCP0932-500G TCP0932-100G
Pentaerythritol tetrakis [3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate] ≥95.0%
Pentaerythritol tetrakis [3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate]

CAS Number: 6683-19-8
MDL Number: MFCD00059345
Molecular Formula: C73H108O12
Molecular Weight: 1177.66
Purity/Analysis Method: >95.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 115
Flash Point (°C): 278


Product Categories: Materials Chemistry, Resin Additives
Other Product Names / Synonyms: Tetrakis[methylene-3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate]methane

Formula: C₇₃H₁₀₈O₁₂
MW: 1177.66 g/mol
Melting Pt: 115 °C
Flash Pt: 278 °C
MDL Number: MFCD00059345
CAS Number: 6683-19-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=80a834aca6b36e5cae4510ac8386248b75f8fdae&vid=80a834aca6b36e5cae4510ac8386248b75f8fdae&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR