5-Dimethylamino-2-(2-pyridylazo)phenol ≥98.0%

Supplier: TCI America

Synonyms: 2-(2-Pyridylazo)-5-dimethylaminophenol

P0910-100MG P0910-1G
TCP0910-100MGEA 148.52 CAD
TCP0910-100MG TCP0910-001G
5-Dimethylamino-2-(2-pyridylazo)phenol ≥98.0%
5-Dimethylamino-2-(2-pyridylazo)phenol

[for Determination of Zinc in Serum]
CAS Number: 50783-80-7
MDL Number: MFCD00059778
Molecular Formula: C13H14N4O
Molecular Weight: 242.28
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: Red
Melting point (°C): 175


Product Categories: Analytical Science, Azo Compounds (Chelating Agents)
Other Product Names / Synonyms: 2-(2-Pyridylazo)-5-dimethylaminophenol

Formula: C₁₃H₁₄N₄O
Melting Pt: 175 °C
MDL Number: MFCD00059778
CAS Number: 50783-80-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c51d3f405a4d8128507f372df4b5289527b98993&vid=c51d3f405a4d8128507f372df4b5289527b98993&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR