2,4-Diaminobenzenesulfonic acid ≥98.0% for synthesis

Supplier: TCI America

Synonyms: 2,4-Diaminobenzenesulphonic acid, 1,3-Phenylenediamine-4-sulfonic acid, 2,4-Diaminobenzenesulfonic acid, 1,3-Phenylenediamine-4-sulphonic acid

P0893-25G P0893-500G
TCP0893-025GEA 67.21 CAD
TCP0893-025G TCP0893-500G
2,4-Diaminobenzenesulfonic acid ≥98.0% for synthesis
2,4-Diaminobenzenesulphonic acid

CAS Number: 88-63-1
MDL Number: MFCD00035772
Molecular Formula: C6H8N2O3S
Molecular Weight: 188.20
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: Slightly Pale Yellow
Flash Point (°C): 300


Other Product Names / Synonyms: 2,4-Diaminobenzenesulfonic Acid

Formula: C₆H₈N₂O₃S
MW: 188.21 g/mol
Boiling Pt: 406
Flash Pt: 300 °C
MDL Number: MFCD00035772
CAS Number: 88-63-1
EINECS: 201-846-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0e73da52e071890326c2529d0c5dd741e30d380f&vid=0e73da52e071890326c2529d0c5dd741e30d380f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR