DMP (Dimethyl pimelimidate dihydrochloride) ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: DMP, Pimelimidic acid dimethyl ester dihydrochloride, Dimethyl pimelimidate dihydrochloride, Dimethyl pimelinediimidate dihydrochloride, Dimethyl heptanebis(imidate) dihydrochloride

P0892-25G P0892-5G
TCP0892-025GEA 636.74 CAD
TCP0892-025G TCP0892-005G
DMP (Dimethyl pimelimidate dihydrochloride) ≥98.0% (by titrimetric analysis)
DMP (Dimethyl pimelimidate dihydrochloride)

[Cross-linking Agent for Peptides Research]
CAS Number: 58537-94-3
MDL Number: MFCD00038942
Molecular Formula: C9H18N2O2
Molecular Weight: 259.17
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Storage Temperature: 0-10°C


Product Categories: Bioscience, Crosslinkers
Other Product Names / Synonyms: Pimelimidic Acid Dimethyl Ester Dihydrochloride

Formula: C₉H₁₈N₂O₂
MW: 259.17 g/mol
MDL Number: MFCD00038942
CAS Number: 58537-94-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=113a7c1d5e1b3b6d65d4ff238b7b9fb1187220ff&vid=113a7c1d5e1b3b6d65d4ff238b7b9fb1187220ff&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR