Sodium 1-pentadecanesulfonate

Supplier: TCI America

P0889-5G P0889-1G
TCP0889-005GEA 1102.52 CAD
TCP0889-005G TCP0889-001G
Sodium 1-pentadecanesulfonate
Sodium 1-pentadecanesulphonate

CAS Number: 5896-54-8
MDL Number: MFCD00059838
Molecular Formula: C15H32O3S
Molecular Weight: 314.46
Form: Crystal
Color: White


Other Product Names / Synonyms: 1-Pentadecanesulfonic Acid Sodium Salt

Formula: C₁₅H₃₁NaO₃S
MDL Number: MFCD00059838
CAS Number: 5896-54-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f953cebe033f8ee91f06f1c3e7cf0ecff9e2a3e0&vid=f953cebe033f8ee91f06f1c3e7cf0ecff9e2a3e0&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR