Pentaerythritol tetrakis(mercaptoacetate) ≥95.0%

Supplier: TCI America

Synonyms: Pentaerythritol Tetrathioglycolate

P0888-25G P0888-500G
TCP0888-025GEA 62.88 CAD
TCP0888-025G TCP0888-500G
Pentaerythritol tetrakis(mercaptoacetate) ≥95.0%
Pentaerythritol tetrakis(mercaptoacetate)

CAS Number: 10193-99-4
MDL Number: MFCD00022080
Molecular Formula: C13H20O8S4
Molecular Weight: 432.54
Purity/Analysis Method: >95.0% (T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 250
Flash Point (°C): 248
Specific Gravity (20/20): 1.39


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Pentaerythritol Tetrathioglycolate

Formula: C₁₃H₂₀O₈S₄
MW: 432.54 g/mol
Boiling Pt: 250 °C
Density: 1.39 (20 °C)
Flash Pt: 248 °C
MDL Number: MFCD00022080
CAS Number: 10193-99-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1e3a568cad02638dfee415f72a3aa92201db3c15&vid=1e3a568cad02638dfee415f72a3aa92201db3c15&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR