Pentaerythritol-tetra(3-mercaptopropionate) ≥90.0% (by titrimetric analysis)
Supplier: TCI America
P0886-25ML
P0886-500ML
TCP0886-025MLEA
63.61
CAD
TCP0886-025ML
TCP0886-500ML
Pentaerythritol-tetra(3-mercaptopropionate) ≥90.0% (by titrimetric analysis)
Pentaerythritol-tetra(3-mercaptopropionate)
CAS Number: 7575-23-7
MDL Number: MFCD00022104
Molecular Formula: C17H28O8S4
Molecular Weight: 488.64
Purity/Analysis Method: >90.0% (T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 275
Flash Point (°C): 250
Specific Gravity (20/20): 1.28
Formula:
C₁₇H₂₈O₈S₄ Boiling Pt: 275 °C Density: 1.28 (20 °C) Flash Pt: 250 °C |
MDL Number:
MFCD00022104 CAS Number: 7575-23-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...