Polyethylene glycol monolaurate, M.W. n=∼10
Supplier: TCI America
P0872-25G
P0872-500G
TCP0872-025GEA
49.82
CAD
TCP0872-025G
TCP0872-500G
Polyethylene glycol monolaurate, M.W. n=∼10
Polyethylene glycol monolaurate
CAS Number: 31943-11-0
MDL Number: MFCD00148005
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 250
Specific Gravity (20/20): 1.04
Other Product Names / Synonyms: Polyethylene Glycol Monododecanoate
Formula:
HO(CH₂CH₂O)nOC(CH₂)₁₀CH₃ Density: 1.04 (20 °C) Flash Pt: 250 °C |
MDL Number:
MFCD00148005 CAS Number: 31943-11-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...