Methyl pentadecanoate ≥98.0%

Supplier: TCI America

P0869-5ML P0869-25ML
TCP0869-5MLEA 143.78 CAD
TCP0869-5ML TCP0869-025ML
Methyl pentadecanoate ≥98.0%
Methyl pentadecanoate

CAS Number: 7132-64-1
MDL Number: MFCD00008989
Molecular Formula: C16H32O2
Molecular Weight: 256.43
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 155
Flash Point (°C): 110
Freezing point (°C): 16
Specific Gravity (20/20): 0.87


Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Pentadecanoic Acid Methyl Ester

Formula: C₁₆H₃₂O₂
Boiling Pt: 155 °C
Density: 0.87 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00008989
CAS Number: 7132-64-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=82a9e387cae3d34049a15297b3238858d004597d&vid=82a9e387cae3d34049a15297b3238858d004597d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR