1H-Pyrazole-1-carboximidamide hydrochloride ≥98.0%

Supplier: TCI America

Synonyms: 1-Amidinopyrazole Hydrochloride, 1H-Pyrazole-1-carboxamidine monohydrochloride, Hydrochloride, 1-Pyrazolecarboxamidine, Praxadine

A2055-25G A2055-5G
TCA2055-25GEA 132.72 CAD
TCA2055-25G TCA2055-5G
1H-Pyrazole-1-carboximidamide hydrochloride ≥98.0%
1H-Pyrazole-1-carboximidamide hydrochloride

CAS Number: 4023-02-3
MDL Number: MFCD00210087
Molecular Formula: C4H6N4
Molecular Weight: 146.58
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 167


Product Categories: Synthetic Organic Chemistry, Guanidinylation
Other Product Names / Synonyms: Praxadine, 1-Pyrazolecarboxamidine Hydrochloride, Pyrazole-1-carboximidamide Hydrochloride, 1-Carbamimidoylpyrazole Hydrochloride

Formula: C₄H₇ClN₄
MW: 146.58 g/mol
Melting Pt: 167 °C
MDL Number: MFCD00210087
CAS Number: 4023-02-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d5f8d795846d4baf2dd4b806466803f14fc05e3&vid=4d5f8d795846d4baf2dd4b806466803f14fc05e3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR