Ethyl pentadecanoate ≥97.0%

Supplier: TCI America

P0868-5ML P0868-25ML
TCP0868-5MLEA 172.22 CAD
TCP0868-5ML TCP0868-025ML
Ethyl pentadecanoate ≥97.0%
Ethyl pentadecanoate

CAS Number: 41114-00-5
MDL Number: MFCD00042872
Molecular Formula: C17H34O2
Molecular Weight: 270.46
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 158
Freezing point (°C): 12
Specific Gravity (20/20): 0.86


Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Pentadecanoic Acid Ethyl Ester

Formula: C₁₇H₃₄O₂
Boiling Pt: 158 °C
Density: 0.86 (20 °C)
MDL Number: MFCD00042872
CAS Number: 41114-00-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f0adf580e48ee1afd02956edeef653d2fc540e9&vid=0f0adf580e48ee1afd02956edeef653d2fc540e9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR