(-)-Perillaldehyde ≥90.0%

Supplier: TCI America

Synonyms: L(-)-Perillaldehyde, L-Perillaldehyde, (-)-4-Isopropenyl-1-cyclohexene-1-carboxaldehyde, (S)-4-(Prop-1-en-2-yl)cyclohex-1-ene-1-carbaldehyde

P0866-25ML P0866-5ML
TCP0866-025MLEA 120.57 CAD
TCP0866-025ML 76220-202
(-)-Perillaldehyde ≥90.0%
(-)-Perillaldehyde

CAS Number: 18031-40-8
MDL Number: MFCD00001543
Molecular Formula: C10H14O
Molecular Weight: 150.22
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Boiling point (°C): 98
Flash Point (°C): 95
Specific Gravity (20/20): 0.97
Specific rotation [a]20/D: -130 deg (neat)


Product Categories: Bioscience, Crude Medicine Ingredients for Research and Experimental Use, Terpenoids (Phytochemicals), Monocyclic Monoterpenes
Other Product Names / Synonyms: (-)-4-Isopropenyl-1-cyclohexene-1-carboxaldehyde

Formula: C₁₀H₁₄O
MW: 150.22 g/mol
Boiling Pt: 98 °C
Density: 0.97 (20 °C)
Flash Pt: 95 °C
MDL Number: MFCD00001543
CAS Number: 18031-40-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a423cef615537f0476002c4382b90337179a92fc&vid=a423cef615537f0476002c4382b90337179a92fc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR