Sodium monododecyl phosphate (mixture of mono and disodium salt)
Supplier: TCI America
P0865-25G
P0865-500G
TCP0865-025GEA
55.3
CAD
TCP0865-025G
TCP0865-500G
Sodium monododecyl phosphate (mixture of mono and disodium salt)
Sodium monododecyl phosphate (mixture of mono and disodium salt)
CAS Number: 7423-32-7
MDL Number: MFCD00058971
Form: Crystal
Color: White
Other Product Names / Synonyms: Phosphoric Acid Monododecyl Ester Sodium Salt, Sodium Monolauryl Phosphate
MDL Number:
MFCD00058971 CAS Number: 7423-32-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...