Phenyl vinyl sulfoxide ≥95.0%

Supplier: TCI America

Synonyms: (Vinylsulfinyl)benzene, (Vinylsulphinyl)benzene

P0863-25G P0863-5G
TCP0863-25GEA 736.28 CAD
TCP0863-25G TCP0863-005G
Phenyl vinyl sulfoxide ≥95.0%
Phenyl vinyl sulfoxide

CAS Number: 20451-53-0
MDL Number: MFCD00002086
Molecular Formula: C8H8OS
Molecular Weight: 152.21
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 110
Flash Point (°C): 110
Specific Gravity (20/20): 1.16

Formula: C₈H₈OS
MW: 152.21 g/mol
Boiling Pt: 110 °C
Density: 1.16 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00002086
CAS Number: 20451-53-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=578692b4d8e623f6e096e70dc7b1b5563377cf6b&vid=578692b4d8e623f6e096e70dc7b1b5563377cf6b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR