2-Phenoxytetrahydropyran ≥90.0%
Supplier: TCI America
P0853-25ML
P0853-5ML
TCP0853-025MLEA
352.34
CAD
TCP0853-025ML
TCP0853-5ML
2-Phenoxytetrahydropyran ≥90.0%
2-Phenoxytetrahydropyran
CAS Number: 4203-50-3
MDL Number: MFCD00059801
Molecular Formula: C11H14O2
Molecular Weight: 178.23
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.08
Other Product Names / Synonyms: Tetrahydro-2-phenoxy-2H-pyran
Formula:
C₁₁H₁₄O₂ Density: 1.08 (20 °C) |
MDL Number:
MFCD00059801 CAS Number: 4203-50-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...